Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxy-5-fluoro-4-pyrimidinone 98.0+%, TCI America™
SDP

Catalog No. E06845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06845G 5 g
E068425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06845G Supplier TCI America Supplier No. E06845G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 56177-80-1
Molecular Formula C6H7FN2O2
Molecular Weight (g/mol) 158.132
InChI Key XREDGJFKPWBNRQ-UHFFFAOYSA-N
Synonym 5-Fluoro-2-ethoxy-4-pyrimidinone
PubChem CID 92041
IUPAC Name 2-ethoxy-5-fluoro-1H-pyrimidin-6-one
SMILES CCOC1=NC=C(C(=O)N1)F

Specifications

Melting Point 180°C
Color White
Quantity 5 g
Formula Weight 158.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethoxy-5-fluoro-4-pyrimidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.