Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxybenzoic Acid, Spectrum™ Chemical
SDP

Catalog No. 18620724 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-724 25 g
18-620-725 100 g
18-620-726 1 kg
18-620-727 5 kg
18-620-728 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-620-724 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E230525GM
Add to Cart
Add to Cart

Synonym: o-Ethoxybenzoic Acid, Benzoic acid, 2-Ethoxy-, Benzoic Acid, o-Ethoxy-

Chemical Identifiers

CAS 134-11-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
InChI Key XDZMPRGFOOFSBL-UHFFFAOYSA-N
IUPAC Name 2-ethoxybenzoic acid
SMILES CCOC1=CC=CC=C1C(O)=O

Specifications

Quantity 25 g
Percent Purity 98%
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.