Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxybenzonitrile 98.0+%, TCI America™
SDP

Catalog No. E096825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E096825G 25 g
E09685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E096825G Supplier TCI America Supplier No. E096825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6609-57-0
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00001789
InChI Key DXTLCLWOCYLDHL-UHFFFAOYSA-N
Synonym o-ethoxybenzonitrile, benzonitrile, ethoxy, 2-ethoxybenzenecarbonitrile, o-ethoxybenzontrle, acmc-20anwu, 2-ethoxy benzonitrile, 2-ethoxy-benzonitrile, pubchem4773, benzonitrile, 2-ethoxy, 2-ethoxybenzonitrile
PubChem CID 576922
IUPAC Name 2-ethoxybenzonitrile
SMILES CCOC1=CC=CC=C1C#N

Specifications

Melting Point 5°C
Color Yellow
Boiling Point 45°C
Quantity 25 g
UN Number 3276
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethoxybenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.