Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxybenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. E049925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E049925G 25 g
E04995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E049925G Supplier TCI America Supplier No. E049925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71672-75-8
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00004616
InChI Key ICJVQAHPHKYCNU-UHFFFAOYSA-N
Synonym 2-ethoxyphenyl methanol, 2-ethoxybenzyl alcohol, benzenemethanol, 2-ethoxy, o-ethoxybenzyl alcohol, ethyl ether of saligenin, 2-ethoxybenzylalcohol, 2-ethoxybenzenemethanol, benzyl alcohol, o-ethoxy, benzenemethanol, 2-ethoxy-9ci, acmc-1bgrn
PubChem CID 51381
IUPAC Name (2-ethoxyphenyl)methanol
SMILES CCOC1=CC=CC=C1CO

Specifications

Color Yellow
Boiling Point 265°C
Quantity 25 g
Formula Weight 152.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethoxybenzyl Alcohol
RTECSNumber DO5467000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.