Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxyethanol, Reagent Grade, 99%, Solstice

Catalog No. 60044758 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-758 2.5 L Glass Bottle
60-044-757 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-758 Supplier Solstice Supplier No. 1280822.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-80-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002869
InChI Key ZNQVEEAIQZEUHB-UHFFFAOYSA-N
Synonym cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
PubChem CID 8076
ChEBI CHEBI:46788
SMILES CCOCCO

Specifications

CAS Max % 100.0000%
Melting Point -90°C (literature)
Density 0.93 g/mL at 25 °C(lit.)
Boiling Point 136°C
Flash Point 40°C
Packaging Glass Bottle
Linear Formula FeCl3
Quantity 2.5 L
UN Number UN1171
Beilstein 1098271
IUPAC Name 2-ethoxyethanol
Formula Weight 90.12
Percent Purity >98.5%
Grade Reagent
Chemical Name or Material 2-Ethoxyethanol, Reagent Grade, 99%
Show More Show Less
ShelfLife 1080 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Ethylene glycol monoethyl ether
EINECSNumber 203-804-1
Recommended Storage Room Temp
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.