Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxyethanol, Spectrum™ Chemical
SDP

Catalog No. 18608210 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-210 4 L Amber Glass Bottle
18-608-207 1 L Amber Glass Bottle
18-608-208 20 L Amber Glass Bottle
18-608-209 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 18-608-210 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E10214LTGL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 110-80-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
InChI Key ZNQVEEAIQZEUHB-UHFFFAOYSA-N
IUPAC Name 2-ethoxyethan-1-ol
SMILES CCOCCO

Specifications

Acidity 5e-05
CAS Max % 1
Boiling Point 135.1°C (275.2°F)
Packaging Amber Glass Bottle
Quantity 4 L
Specific Gravity 0.9290 - 0.9330
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethanol
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.