Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxynaphthalene 98.0+%, TCI America™
SDP

Catalog No. N043125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N043125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N043125G Supplier TCI America Supplier No. N043125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 93-18-5
Melting Point 35°C
Color White
Boiling Point 282°C
Molecular Formula C12H12O
Quantity 25 g
MDL Number MFCD00016808
Synonym bromelia, naphthalene, 2-ethoxy, neroline, nerolin new, nerolin ii, ethyl 2-naphthyl ether, 2-naphthol ethyl ether, bromelia compound, beta-naphthyl ethyl ether, ethyl beta-naphthyl ether
InChI Key GUMOJENFFHZAFP-UHFFFAOYSA-N
SMILES CCOC1=CC2=CC=CC=C2C=C1
IUPAC Name 2-ethoxynaphthalene
Molecular Weight (g/mol) 172.227
PubChem CID 7129
Formula Weight 172.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethoxynaphthalene
Show More Show Less
EINECSNumber (4)-0362
RTECSNumber QJ6900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.