Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxypyrazine 98.0+%, TCI America™
SDP

Catalog No. E06975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06975G 5 g
E069725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06975G Supplier TCI America Supplier No. E06975G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 38028-67-0
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD00014609
InChI Key ATIUKJHILQVQLE-UHFFFAOYSA-N
Synonym pyrazine, ethoxy, 2-ethoxy pyrazine, pyrazine, 2-ethoxy, 3-ethoxypyrazine, 2-ethoxy-pyrazine, acmc-1bmsg, ksc222g4b, chembl93554
PubChem CID 2758778
IUPAC Name 2-ethoxypyrazine
SMILES CCOC1=NC=CN=C1

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 g
UN Number 1993
Formula Weight 124.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethoxypyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.