Learn More
Sigma Organic Chemistry 2-Ethyl-1 3-hexanediol | 500G | 94-96-2 | MFCD00004578
Supplier: Sigma Organic Chemistry E29125500G

2-Ethyl-1,3-hexanediol, 500G
About This Item:
Synonym(s): Ethylhexylene glycol
Linear Formula: CH3CH2CH2CH(OH)CH(C2H5)CH2OH
Molecular Weight: 146.23
CAS Number: 94-96-2
MDL Number: MFCD00004578
UNSPSC Code: 12352100
Purity: 0.97
Boiling Point: 241-249 C (lit.)
Melting Point: -40 C (lit.)
Density: 0.933 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.451 (lit.)
Storage: room temp
EINECS Number: 202-377-9
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.