Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-1-butanol 98.0+%, TCI America™
SDP

Catalog No. E0068100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0068100ML 100 mL
E006825ML 25 mL
E0068500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0068100ML Supplier TCI America Supplier No. E0068100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 97-95-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00004744
InChI Key TZYRSLHNPKPEFV-UHFFFAOYSA-N
Synonym 2-ethyl-1-butanol, 1-butanol, 2-ethyl, 2-ethylbutanol, 2-ethylbutyl alcohol, pseudohexyl alcohol, 3-methylolpentane, 2-ethylbutanol-1, ethylbutanol, 3-pentylcarbinol, sec-pentylcarbinol
PubChem CID 7358
IUPAC Name 2-ethylbutan-1-ol
SMILES CCC(CC)CO

Specifications

Melting Point -15°C
Color Colorless
Boiling Point 146°C
Quantity 100 mL
UN Number 2275
Formula Weight 102.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-1-butanol
EINECSNumber (2)-0217
RTECSNumber EL3850000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.