Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-1-hexanol, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019155 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9155 5 mL
11-102-0585 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9155 Supplier MilliporeSigma Supelco Supplier No. 086075MLF
Add to Cart
Add to Cart

2-Ethyl-1-hexanol is an industrial chemical widely used in the production of polyvinyl chloride(PVC) plasticizers and is a potent indoor air pollutant emitted mainly from furniture or carpets with a PVC backing.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 104-76-7
Ignition Point 550°F
Vapor Pressure 0.2 mmHg (20°C)
Vapor Density 4.49 (vs. air)
Density 0.833 g/mL (at 25°C)
Flash Point 75°C - closed cup
Linear Formula CH3(CH2)3 CH(C2H5)CH2 OH
MDL Number MFCD00004746
Melting Point -76°C
Boiling Point 183°C to 186°C
Synonym Isooctyl alcohol
Type Analytical Standard
Quantity 5 mL
Formula Weight 130.23
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.