Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-3-(methylthio)pyrazine 98.0+%, TCI America™
SDP

Catalog No. E071925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E071925G 25 g
E07195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E071925G Supplier TCI America Supplier No. E071925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 72987-62-3
Molecular Formula C7H10N2S
Molecular Weight (g/mol) 154.231
MDL Number MFCD00038027
InChI Key XYHPPOMSLGJAAM-UHFFFAOYSA-N
Synonym 2-(Methylthio)-3-ethylpyrazine
PubChem CID 175366
IUPAC Name 2-ethyl-3-methylsulfanylpyrazine
SMILES CCC1=NC=CN=C1SC

Specifications

Quantity 25 g
Formula Weight 154.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-3-(methylthio)pyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.