Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylbutyryl chloride, 99%

Catalog No. AC409820050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC409820050 5 g Glass bottle
AC409820250 25 g Glass bottle
AC409821000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC409820050 Supplier Thermo Scientific Chemicals Supplier No. 409820050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2736-40-5 | C6H11ClO | 134.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2736-40-5
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.60
MDL Number MFCD00018812
InChI Key SMUKODJVMQOSAB-UHFFFAOYSA-N
Synonym 2-ethylbutyryl chloride, 2-ethylbutyroyl chloride, butanoyl chloride, 2-ethyl, butyryl chloride, 2-ethyl, 2-ethylbutyrylchloride, diethylacetylchloride, diethylacetyl chloride, acmc-1cqal, 2-ethyl butyryl chloride, 2-ethyl-butyryl chloride
PubChem CID 75954
IUPAC Name 2-ethylbutanoyl chloride
SMILES CCC(CC)C(Cl)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9820g/mL
Boiling Point 138°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4230 to 1.425
Linear Formula (C2H5)2CHCOCl
Quantity 5 g
Specific Gravity 0.982
Formula Weight 134.61
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Ethylbutyryl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 220-359-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.