Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexyl 2-Cyano-3,3-diphenylacrylate 98.0+%, TCI America™
SDP

Catalog No. C0968100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0968100G 100 g
C096825G 25 g
C0968500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0968100G Supplier TCI America Supplier No. C0968100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6197-30-4
Molecular Formula C24H27NO2
Molecular Weight (g/mol) 361.49
MDL Number MFCD00059260
InChI Key FMJSMJQBSVNSBF-UHFFFAOYNA-N
Synonym 2-Cyano-3,3-diphenylacrylic Acid 2-Ethylhexyl Ester, Octyl 2-Cyano-3,3-diphenylacrylate
PubChem CID 22571
IUPAC Name 2-ethylhexyl 2-cyano-3,3-diphenylprop-2-enoate
SMILES CCCCC(CC)COC(=O)C(C#N)=C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -10°C
Color Yellow
Boiling Point 218°C
Quantity 100 g
Formula Weight 361.49
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl 2-Cyano-3,3-diphenylacrylate
EINECSNumber (4)-0262
RTECSNumber UD3328750
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.