Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexyl 4-(Dimethylamino)benzoate 98.0+%, TCI America™
SDP

Catalog No. D1871100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1871100ML 100 mL
D187125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1871100ML Supplier TCI America Supplier No. D1871100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21245-02-3
Molecular Formula C17H27NO2
Molecular Weight (g/mol) 277.408
MDL Number MFCD00017526
InChI Key WYWZRNAHINYAEF-UHFFFAOYSA-N
Synonym 2-ethylhexyl 4-dimethylamino benzoate, padimate o, escalol 507, padimate-o, octyl dimethyl paba, arlatone uvb, benzoic acid, 4-dimethylamino-, 2-ethylhexyl ester, 2-ethylhexyl p-dimethylaminobenzoate, eusolex 6007, sundown
PubChem CID 30541
IUPAC Name 2-ethylhexyl 4-(dimethylamino)benzoate
SMILES CCCCC(CC)COC(=O)C1=CC=C(C=C1)N(C)C

Specifications

Color Yellow
Boiling Point 325°C
Quantity 100 mL
Formula Weight 277.41
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl 4-(Dimethylamino)benzoate
EINECSNumber (3)-1436
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.