Learn More
Sigma Organic Chemistry 2-Ethylhexyl benzoate | 1G | 5444-75-7 | MFCD00072620
Supplier: Sigma Organic Chemistry CDS0005331G

2-Ethylhexyl benzoate , 1G
About this Item:
CAS #: 5444-75-7
MDL #: MFCD00072620
Molecular Weight: 234.33
UNSPSC Code: 12352200
Chemical Formula: C15H22O2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.