Learn More
2-Ethylhexyl bromide, 96%, stab. with copper
Description
2-Ethylhexyl bromide is used in the synthesis of methoxy-4-(2-ethylhexyloxy)benzene, 1-bromo-4-(2?-ethylhexyloxy)benzene and 3,6-diaryl substituted 9-alkylcarbazoles as hole transporting materials for various organic light emitting devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Ethylhexyl bromide is used in the synthesis of methoxy-4-(2-ethylhexyloxy)benzene, 1-bromo-4-(2′-ethylhexyloxy)benzene and 3,6-diaryl substituted 9-alkylcarbazoles as hole transporting materials for various organic light emitting devices.
Solubility
Soluble in chloroform, ethyl acetate. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat, oxidizing agents.
Specifications
Specifications
| Density | 1.086 |
| Boiling Point | 94°C to 95°C (35 mmHg) |
| Flash Point | 69°C (156°F) |
| Refractive Index | 1.4538 |
| Quantity | 100 g |
| Beilstein | 1098313 |
| Solubility Information | Soluble in chloroform,ethyl acetate. Insoluble in water. |
| Formula Weight | 193.13 |
| Percent Purity | 96% |
| Chemical Name or Material | 2-Ethylhexyl bromide, stabilized with copper |
Safety and Handling
| Hazard Category | H227-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| EINECSNumber | 242-659-9 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.