Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexyl Phosphate (Mono- and Di- Ester mixture), TCI America™
SDP

Catalog No. P026125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P026125G 25 g
P0261500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P026125G Supplier TCI America Supplier No. P026125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 12645-31-7
MDL Number MFCD00135884
Synonym Phosphoric Acid 2-Ethylhexyl Ester, Octyl Phosphate, Phosphoric Acid Octyl Ester

Specifications

Color Yellow
Quantity 25 g
UN Number 3265
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl Phosphate (Mono- and Di- Ester mixture)
EINECSNumber (2)-1986
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.