Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexyl Thioglycolate 98.0+%, TCI America™
SDP

Catalog No. T061225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T061225ML 25 mL
T0612500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T061225ML Supplier TCI America Supplier No. T061225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7659-86-1
Molecular Formula C10H20O2S
Molecular Weight (g/mol) 204.33
MDL Number MFCD00022081
InChI Key OWHSTLLOZWTNTQ-UHFFFAOYNA-N
Synonym Thioglycolic Acid 2-Ethylhexyl Ester, Mercaptoacetic Acid 2-Ethylhexyl Ester, Octyl Thioglycolate, Thioglycolic Acid Octyl Ester, 2-Ethylhexyl Mercaptoacetate
PubChem CID 24309
IUPAC Name 2-ethylhexyl 2-sulfanylacetate
SMILES CCCCC(CC)COC(=O)CS

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 204.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl Thioglycolate
EINECSNumber (2)-1359
RTECSNumber AI7255000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.