Learn More
2-Ethylimidazole, 98%
Description
2-Ethylimidazole is used as internal standard in the quantification of 4-(5-)methylimidazole (4MI) in coffee by GC-MS method. It is a versatile intermediate used as a building block for active ingredients as well as in epoxy curing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Ethylimidazole is used as internal standard in the quantification of 4-(5-)methylimidazole (4MI) in coffee by GC-MS method. It is a versatile intermediate used as a building block for active ingredients as well as in epoxy curing.
Solubility
Soluble in water (617 g/L at 20°C)
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 78°C to 86°C |
| Boiling Point | 268°C |
| Flash Point | 159°C (318°F) |
| Quantity | 100 g |
| Beilstein | 106381 |
| Solubility Information | Soluble in water (617g/L at 20°C) |
| Formula Weight | 96.13 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Ethylimidazole |
Safety and Handling
| Hazard Category | H302-H315-H318-H335 |
| Hazard Statement | GHS H Statement H318-H302-H335-H315 Causes serious eye damage. Harmful if swallowed. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | P234-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P390-P501c |
| EINECSNumber | 214-011-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.