Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylpiperidine 95%, Thermo Scientific™

Catalog No. AC118891000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC118891000 100 mL
AC118895000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC118891000 Supplier Thermo Scientific Chemicals Supplier No. 118891000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1484-80-6
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.204
InChI Key QBBKKFZGCDJDQK-UHFFFAOYSA-N
Synonym piperidine, 2-ethyl, .alpha.-ethylpiperidine, 2-ethylpiperidin, 2-ethyl-piperidine, alpha-ethylpiperidine, 2-ethvlpiperidine, 2-ethyl-piperidin, 2-ethyl piperidine, pubchem6758, acmc-1bgny
PubChem CID 94205
IUPAC Name 2-ethylpiperidine
SMILES CCC1CCCCN1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.87
Infrared Spectrum Authentic
Refractive Index 1.4500 to 1.4520 (20°C, 589nm)
Quantity 100 mL
Beilstein 20, 104
Formula Weight 113.2
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material 2-Ethylpiperidine, 95%
Show More Show Less
EINECSNumber 216-058-7
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.