Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Ethylsulfonyl)ethanol, 94%, Thermo Scientific™

Catalog No. AAB2018006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2018006 5 g
AAB2018014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2018006 Supplier Thermo Scientific Chemicals Supplier No. B2018006

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-12-2
Molecular Formula C4H10O3S
Molecular Weight (g/mol) 138.181
MDL Number MFCD00041259
InChI Key LJNDRJPZYNGSKZ-UHFFFAOYSA-N
Synonym 2-ethylsulfonyl ethanol, ethylsulfonylethanol, ethanol, 2-ethylsulfonyl, ethylsulfonylethyl alcohol, ethyl 2-hydroxyethyl sulfone, unii-95yyw1049k, 2-ethanesulfonyl ethanol, ese, beta-ethylsulfonylethanol, 2-ethanesulfonyl-ethanol
PubChem CID 10549
IUPAC Name 2-ethylsulfonylethanol
SMILES CCS(=O)(=O)CCO

Specifications

Melting Point 35°C to 41°C
Density 1.25
Boiling Point 153°C (2.5 mmHg)
Flash Point 188°C (370°F)
Quantity 5 g
Beilstein 1748754
Sensitivity Hygroscopic
Formula Weight 138.19
Percent Purity 94%
Chemical Name or Material 2-(Ethylsulfonyl)ethanol
EINECSNumber 000-000-0
RTECSNumber KL1100000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.