Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Ethylthio)ethanol, 96%, Thermo Scientific™

Catalog No. AAL0504018
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0504018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0504018 Supplier Thermo Scientific Chemicals Supplier No. L0504018

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-77-0
Molecular Formula C4H10OS
Molecular Weight (g/mol) 106.18
MDL Number MFCD00002909
InChI Key LNRIEBFNWGMXKP-UHFFFAOYSA-N
Synonym 2-ethylthio ethanol, ethyl 2-hydroxyethyl sulfide, ethanol, 2-ethylthio, ethyl thioethanol, 2-hydroxyethyl ethyl sulfide, ethyl 2-hydroxyethyl thioether, beta-ethylthioethanol, 2-ethylsulfanyl ethanol, beta-hydroxydiethyl sulfide, 2-ethylthioethanol
PubChem CID 8075
IUPAC Name 2-ethylsulfanylethanol
SMILES CCSCCO

Specifications

Melting Point ∼-100°C
Density 1.018
Boiling Point 180°C to 184°C
Flash Point 93°C (199°F)
Refractive Index 1.4869
Quantity 50 g
UN Number UN1760
Beilstein 1731373
Merck Index 14,3857
Formula Weight 106.19
Percent Purity 96%
Chemical Name or Material 2-(Ethylthio)ethanol
Show More Show Less
missing translation for 'healthHazard1' H227-H318
missing translation for 'healthHazard2' GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
missing translation for 'healthHazard3' P210-P280-P305+P351+P338-P310-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 203-802-0
RTECSNumber KL1225000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.