Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-3-iodopyridine 98.0+%, TCI America™
SDP

Catalog No. F07331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07331G 1 g
F07335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07331G Supplier TCI America Supplier No. F07331G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 113975-22-7
Molecular Formula C5H3FIN
Molecular Weight (g/mol) 222.989
MDL Number MFCD03095287
InChI Key WCDCAXVNBOLWNO-UHFFFAOYSA-N
Synonym 2-fluoro-3-iodo-pyridine, pyridine, 2-fluoro-3-iodo, zlchem 409, 2-fluoro-3 iodopyridine, 2-fluoro-3-iodo-pyridin, 2-fluoro-3-iodopyridine, 2-fluoro-3-iodopyridine;, 2-fluoro-3-iodo pyridine, acmc-1c73w, ksc174s0n
PubChem CID 7009501
IUPAC Name 2-fluoro-3-iodopyridine
SMILES C1=CC(=C(N=C1)F)I

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 99°C
Quantity 1 g
Formula Weight 222.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-3-iodopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.