Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-biphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F07851G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07851G 1 g
F0785-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07851G Supplier TCI America Supplier No. F07851G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 178305-99-2
Molecular Formula C12H10BFO2
MDL Number MFCD01075707
InChI Key BWYWXDFYJSIUBE-UHFFFAOYSA-N
PubChem CID 2774698

Specifications

Melting Point 248°C
Color White-Yellow
Quantity 1 g
Synonym 2-fluoro-4-biphenylylboronic acid, 2-fluoro-4-biphenylboronic acid, 2-fluorobiphenyl-4-boronic acid, 2-fluorobiphenyl-4-ylboronic acid, 3-fluoro-4-phenylphenyl boranediol, 3-fluoro-4-phenylbenzeneboronic acid, 2-fluoro 1,1'-biphenyl-4-boronic acid, 3-fluoro-4-phenyl phenylboronic acid, 2-fluoro-1,1'-biphenyl-4-yl boronic acid, 2-fluoro-1,1'-biphenyl-4-boronic acid
SMILES B(C1=CC(=C(C=C1)C2=CC=CC=C2)F)(O)O
IUPAC Name (3-fluoro-4-phenylphenyl)boronic acid
Molecular Weight (g/mol) 216.018
Formula Weight 216.02
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-4-biphenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.