Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-methylaniline 97.0+%, TCI America™
SDP

Catalog No. F052925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F052925G 25 g
F05295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F052925G Supplier TCI America Supplier No. F052925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 452-80-2
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00040975
InChI Key ZQEXBVHABAJPHJ-UHFFFAOYSA-N
Synonym 2-fluoro-4-methyaniline, 2-fluoro-p-toluidine, 4-amino-3-fluorotoluene, p-toluidine, 2-fluoro, benzenamine, 2-fluoro-4-methyl, 2-fluoro-4-methyl aniline, 2-fluoro-4-methyl-aniline, 2-fluoro-4-methyl-phenylamine, 2-fluoro-4-methylphenylamine, benzenamine, 2-fluoro-4-methyl-9ci
PubChem CID 67984
IUPAC Name 2-fluoro-4-methylaniline
SMILES CC1=CC=C(N)C(F)=C1

Specifications

Melting Point 3°C
Boiling Point 71°C
Quantity 25 g
UN Number 2810
Formula Weight 125.15
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-4-methylaniline
RTECSNumber XU6650000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.