Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F06831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06831G 1 g
F0683-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06831G Supplier TCI America Supplier No. F06831G
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 2-Fluoro-4-methylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 170981-26-7
Melting Point 230°C
Color White-Yellow
Molecular Formula C7H8BFO2
Quantity 1 g
MDL Number MFCD05664239
Synonym 2-fluoro-4-methylphenyl boronic acid, 2-fluoro-4-methylbenzeneboronic acid, 2-fluoro-4-methyl-phenyl boronic acid, boronic acid, 2-fluoro-4-methylphenyl, 2-fluoro-p-tolylboronic acid, 4-methyl-2-fluorophenylboronic acid, boronic acid, b-2-fluoro-4-methylphenyl, pubchem23775, ksc489k8t, 2-fluoro-4methylphenylboronic acid
InChI Key LIXXGOMAGHXIMP-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)C)F)(O)O
IUPAC Name (2-fluoro-4-methylphenyl)boronic acid
Molecular Weight (g/mol) 153.947
PubChem CID 2783132
Formula Weight 153.95
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.