Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-nitroaniline, 98%, Thermo Scientific™

Catalog No. AC457601000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457601000 100 g
AC457600050 5 g
AC457600250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC457601000 Supplier Thermo Scientific Chemicals Supplier No. 457601000

Chemical Identifiers

CAS 369-35-7
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.11
InChI Key LETNCFZQCNCACQ-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitrobenzenamine, 2-fluoro-4-nitro-phenylamine, benzenamine, 2-fluoro-4-nitro, 2-fluoro-4-nitrophenylamine, 4-amino-3-fluoronitrobenzene, 2-fluoro-4-nitroaniline, pubchem8503, acmc-1bncp, 2-fluoro-4-niroaniline, 2-fluoro-4-nitro-aniline
PubChem CID 101254
IUPAC Name 2-fluoro-4-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])F)N

Specifications

Melting Point 122.0°C to 130.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula FC6H3(NO2)NH2
Quantity 100 g
Formula Weight 156.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-nitroaniline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position
EINECSNumber 206-719-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.