Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-6-(trifluoromethyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. F082625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F082625G 25 g
F08265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F082625G Supplier TCI America Supplier No. F082625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 94239-04-0
Molecular Formula C6H3F4N
Molecular Weight (g/mol) 165.091
MDL Number MFCD03092901
InChI Key IZOIOCQPMHHDHN-UHFFFAOYSA-N
Synonym 2-fluoro-6-trifluoromethyl pyridine, 2-trifluoromethyl-6-fluoropyridine, pyridine, 2-fluoro-6-trifluoromethyl, 6-fluoro-2-trifluoromethyl pyridine, pubchem2403, acmc-209rqu, ksc488k3d, tpc-py087, 2-fluoro-6-trifluoromethyl py, 2-fluoro-6-rifluoromethylpyridine
PubChem CID 2736501
IUPAC Name 2-fluoro-6-(trifluoromethyl)pyridine
SMILES C1=CC(=NC(=C1)F)C(F)(F)F

Specifications

Color Colorless
Quantity 25 g
UN Number 1993
Formula Weight 165.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.