Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-9-beta-D-arabinofuranosyladenine, 98%

Catalog No. AAJ6556203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6556203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6556203 Supplier Thermo Scientific Chemicals Supplier No. J6556203
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 21679-14-1 | C10H12FN5O4 | 285.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21679-14-1
Molecular Formula C10H12FN5O4
Molecular Weight (g/mol) 285.24
MDL Number MFCD00132942
InChI Key HBUBKKRHXORPQB-FJFJXFQQSA-N
Synonym fludarabine, f-ara-a, 2-fluoro ara-a, 9-beta-d-arabinofuranosyl-2-fluoroadenine, fludarabine inn, fludarabinum latin, fludarabina spanish, fludarabina, fludarabinum, 2r,3s,4s,5r-2-6-amino-2-fluoro-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol
PubChem CID 657237
IUPAC Name (2R,3S,4S,5R)-2-(6-amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O)C2=NC(F)=N1

Specifications

Color White to Yellow
Quantity 1 g
Beilstein 1225932
Merck Index 14,4126
Solubility Information Soluble in DMF, DMSO, methanol or ethanol; Sparingly soluble in water
Formula Weight 285.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Fluoro-9-beta-D-arabinofuranosyladenine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 244-525-5
RTECSNumber AU6207000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.