Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoroadenosine 96.0+%, TCI America™
SDP

Catalog No. F0656200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0656200MG 200 mg
F06561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0656200MG Supplier TCI America Supplier No. F0656200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 146-78-1
Molecular Formula C10H12FN5O4
Molecular Weight (g/mol) 285.24
MDL Number MFCD00866394
InChI Key HBUBKKRHXORPQB-SBTUYZBDNA-N
Synonym 2-fluoroadenosine, adenosine, 2-fluoro, 2r,3r,4s,5r-2-6-amino-2-fluoro-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 6-amino-2-fluoro-9-beta-d-ribofuranosylpurine, 2-far, 2-fas, unii-0s67290crj, 2-6-amino-2-fluoro-purin-9-yl-5-hydroxymethyl-tetrahydro-furan-3,4-diol, purine, 6-amino-2-fluoro-9-beta-d-ribofuranosyl, 2'-fluoroadenosine
PubChem CID 8975
ChEBI CHEBI:39740
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC(F)=N1

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 200 mg
Formula Weight 285.24
Percent Purity ≥96.0% (HPLC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoroadenosine
RTECSNumber AU7386000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.