Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. F055725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F055725G 25 g
F05575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F055725G Supplier TCI America Supplier No. F055725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 30058-40-3
Molecular Formula C6H6FNO2S
Molecular Weight (g/mol) 175.18
MDL Number MFCD00042283
InChI Key WFLBWYLZCQOPCA-UHFFFAOYSA-N
Synonym 2-fluorobenzene-1-sulfonamide, benzenesulfonamide, 2-fluoro, 2-fbsa, 2weg, pubchem11761, acmc-209hcr, 2-fluorophenylsulfonamide, 2-sulfamoyl fluorobenzene, 2-fluorobenzenesulphonamide, 2-flourobenzene sulfonamide
PubChem CID 193663
IUPAC Name 2-fluorobenzene-1-sulfonamide
SMILES NS(=O)(=O)C1=CC=CC=C1F

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 25 g
Formula Weight 175.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluorobenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.