Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzyl Cyanide 97.0+%, TCI America™
SDP

Catalog No. F025725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F025725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F025725G Supplier TCI America Supplier No. F025725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 326-62-5
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD00001897
InChI Key DAVJMKMVLKOQQC-UHFFFAOYSA-N
Synonym 2-fluorophenylacetonitrile, 2-fluorobenzyl cyanide, 2-2-fluorophenyl acetonitrile, benzeneacetonitrile, 2-fluoro, o-fluorophenylacetonitrile, o-fluorobenzyl cyanide, 2-fluorophenyl acetonitrile, acetonitrile, o-fluorophenyl, 2-2-fluorophenyl ethanenitrile, 2-fluorobenzeneacetonitrile
PubChem CID 67592
IUPAC Name 2-(2-fluorophenyl)acetonitrile
SMILES FC1=CC=CC=C1CC#N

Specifications

Boiling Point 235°C
Quantity 25 g
UN Number 1992
Formula Weight 135.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzyl Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.