Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobiphenyl Reference Material, ISO 17034, Quantitative, 1 mL

Catalog No. USATS1401
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-ATS-140-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-ATS-140-1 Supplier Agilent Technologies Supplier No. ATS1401
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 321-60-8 , 75-09-2
Molecular Formula C12H9F
Molecular Weight (g/mol) 172.20
MDL Number MFCD00000317
InChI Key KLECYOQFQXJYBC-UHFFFAOYSA-N
PubChem CID 67579
IUPAC Name 2-fluoro-1,1'-biphenyl
SMILES FC1=CC=CC=C1C1=CC=CC=C1

Specifications

CAS Max % 99.8, 0.15
Analyte or Component Names 2-Fluorobiphenyl,methylene chloride
Element 2-Fluorobiphenyl, Methylene Chloride
Solvent or Matrix Methylene Chloride
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 2000μg/mL
Chemical Name or Material 2-Fluorobiphenyl Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.