Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Fluorophenyl)thiourea 98.0+%, TCI America™
SDP

Catalog No. F10551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10551G 1 g
F10555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10551G Supplier TCI America Supplier No. F10551G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 656-32-6
Molecular Formula C7H7FN2S
Molecular Weight (g/mol) 170.205
MDL Number MFCD00041148
InChI Key WYVZQQOFMQRNPF-UHFFFAOYSA-N
Synonym 1-2-fluorophenyl thiourea, 2-fluorophenyl thiourea, 1-2-fluorophenyl-2-thiourea, thiourea, 2-fluorophenyl, n-2-fluorophenyl thiourea, amino 2-fluorophenyl amino methane-1-thione, maybridge1_008774, acmc-1b7vs, 2-fluoro-phenyl-thiourea
PubChem CID 2737500
IUPAC Name (2-fluorophenyl)thiourea
SMILES C1=CC=C(C(=C1)NC(=S)N)F

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 170.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Fluorophenyl)thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.