Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorophenylacetone 97.0+%, TCI America™
SDP

Catalog No. F097025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F097025G 25 g
F09705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F097025G Supplier TCI America Supplier No. F097025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2836-82-0
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.168
MDL Number MFCD00000325
InChI Key BANVZEUCJHUPOI-UHFFFAOYSA-N
Synonym 2-fluorophenylacetone, 1-2-fluorophenyl propan-2-one, 1-2-fluorophenyl acetone, 2-fluorophenyl acetone, o-fluorophenylacetone, 2-propanone, 1-2-fluorophenyl, acmc-20anub, ksc497e4p, 1-2-fluorophenyl acetone #
PubChem CID 76086
IUPAC Name 1-(2-fluorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=CC=C1F

Specifications

Color Yellow
Boiling Point 47°C
Quantity 25 g
Formula Weight 152.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluorophenylacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.