Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoropyridine-4-methanol, 97%

Catalog No. p-4812812
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452370010 1 g
AC452370050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC452370010 Supplier Thermo Scientific Chemicals Supplier No. 452370010
Only null left
Add to Cart

CAS: 131747-60-9 | C6H6FNO | 127.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131747-60-9
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.12
InChI Key IAFXOPAZVVMMLE-UHFFFAOYSA-N
Synonym 2-fluoropyridin-4-yl methanol, 2-fluoro-4-pyridinemethanol, 2-fluoro-pyridin-4-yl-methanol, 4-pyridinemethanol, 2-fluoro, acmc-1c3lv, 2-fluoropyridine-4-methanol, 2-fluoro-4-hydroxymethylpyridine, 4-pyridinemethanol, 2-fluoro-9ci
PubChem CID 11332468
IUPAC Name (2-fluoropyridin-4-yl)methanol
SMILES C1=CN=C(C=C1CO)F

Specifications

Melting Point 61°C to 63°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 127.12
Percent Purity 97%
Chemical Name or Material 2-Fluoropyridine-4-methanol
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.