Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Formylbenzenesulfonic acid sodium salt

Catalog No. 12050100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-050-100G 100 g
12-050-500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-050-100G Supplier Sigma Aldrich Supplier No. 12050100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Formylbenzenesulfonic acid sodium salt reacts with chitosan in the presence of sodium cyanoborohydride to yield N-benzyl derivatives.2-Formylbenzenesulfonic acid sodium salt was used as precursor to test the ability of fungal strains for transformation of phenolic and non-phenolic precursors into stable and non-toxic dyes.

Specifications

CAS 1008-72-6
Molecular Formula C7H5NaO4S
Linear Formula C7H5NaO4S
MDL Number MFCD00007478
Quantity 100 g
Synonym 2-Sulfobenzaldehyde sodium salt; Benzaldehyde-2-sulfonic acid sodium salt; Sodium 2-formyl-benzolsulfonate
Molecular Weight (g/mol) 208.17
Percent Purity ≥95.0% (T)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.