Learn More
Sigma Organic Chemistry 2-furfuryl isothiocyan | 250mg | 4650-60-6 | MFCD00014112
Supplier: Sigma Organic Chemistry CDS003797250MG

2-furfuryl isothiocyan , 250mg
About this Item:
CAS #: 4650-60-6
MDL #: MFCD00014112
Molecular Weight: 139.18
UNSPSC Code: 12352200
Chemical Formula: C6H5NOS
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.