Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Furonitrile, 99%

Catalog No. p-137269
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170430050 5 g Glass bottle
AC170430250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170430050 Supplier Thermo Scientific Chemicals Supplier No. 170430050
Only null left
Add to Cart

CAS: 617-90-3 | C5H3NO | 93.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-90-3
Molecular Formula C5H3NO
Molecular Weight (g/mol) 93.09
MDL Number MFCD00003223
InChI Key YXDXXGXWFJCXEB-UHFFFAOYSA-N
Synonym 2-furonitrile, 2-cyanofuran, 2-furancarbonitrile, 2-furyl cyanide, unii-2lrk86h722, .alpha.-furyl cyanide, pubchem6961, acmc-209mwb, 2-cyanofuran;2-furancarbonitrile
PubChem CID 69245
IUPAC Name furan-2-carbonitrile
SMILES N#CC1=CC=CO1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow to Orange
Density 1.0600g/mL
Boiling Point 146.0°C to 148.0°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4780 to 1.4800
Quantity 5 g
Beilstein 18, 278
Specific Gravity 1.06
Solubility Information Solubility in water: slightly. Other solubilities: soluble in most common organic solvents
Formula Weight 93.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Furonitrile, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 210-537-4
TSCA TSCA
Recommended Storage Product may darken on storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.