Learn More
Sterilis Solutions 2 Gallon Sharps Container
SDP
Catalog No. 23777028
Change view
Click to view available options
Quantity:
20/Cs.
40/Cs.
60/Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-777-028 20/Cs.
23-777-029 40/Cs.
23-777-030 60/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-777-028 Supplier Sterilis Solutions Supplier No. 3823343701
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sterilis Solutions 2 Gallon Sharps Container - Puncture-resistant sharps container with wide mailbox opening for easy loading of needles.
  • Used to comply with FDA and OSHA standards for sharps and biohazard waste collection.
  • General-purpose sharps container with clear locking lid and marked with biohazard label.
  • Puncture-resistant with wide mailbox slot opening for horizontal loading of needles.
  • Made from transparent red puncture-resistant polypropylene that can be autoclaved or incinerated.
  • Rotating handle for easy transporting and handling.
  • Nests to conserve storage space
Specifications
Certifications/Compliance ISO23907-1:2019 and ISO23908
Closure Type Mailbox Lid
Closure Color White/Translucent
Capacity (English) 2 gal.
Color Red
Dimensions (L x W x H) 13.3 x 11.4 x 5.8 in.
Disposable Single-use
Latex Free Yes
Quantity 20/Cs.
Sterility Non-sterile
Autoclavable Autoclavable
Material Polypropylene
Product Type Sharps Container