Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Heptadecylimidazole, Spectrum™ Chemical
SDP

Catalog No. 18620810 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-810 25 g
18-620-811 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-810 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H127525GM
Add to Cart
Add to Cart

Synonym: 2-Heptadecyl-1H-Imidazole, 1H-Imidazole, 2-Heptadecyl-

Chemical Identifiers

CAS 23328-87-2

Specifications

Packaging Amber Bottle
Quantity 25 g
Molecular Weight (g/mol) 306.54
Percent Purity 85.0%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material 2-Heptadecylimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.