Learn More
2-Heptanone, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | -35.0°C |
| Color | Colorless |
| Density | 0.8200g/mL |
| Boiling Point | 149.0°C to 150.0°C |
| Flash Point | 39°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Plastic drum |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Warning |
| missing translation for 'healthHazard2' | GHS H Statement Harmful if inhaled. Harmful if swallowed. May cause drowsiness or dizziness. Flammable liquid and vapor. |
| missing translation for 'healthHazard3' | GHS P Statement IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF SWALLOWED: rinse mouth. Do NOT induce vomi |
| EINECSNumber | 203-767-1 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.