Learn More
Abcam 2-Heptyl-3-hydroxy-4-quinolone, Intercellular Signal Compound, Solid, 25 mg
Description
- Molecular weight: 259.34 g/mol
- Purity: >96%
- Form: Solid
- Composition: 100% 2-Heptyl-3-hydroxy-4(1H)-quinolinone
- Function: Quorum sensing signaling molecule used to induce and study the regulation of virulence genes
- Storage: Store at ambient temperature
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | 2-Heptyl-3-hydroxy-4-quinolone |
| Shipping Condition | Ambient |
| Form | Solid |
| Buffer | 100% 2-Heptyl-3-hydroxy-4(1H)-quinolinone |
| Content And Storage | Store at ambient temperature. |
| Quantity | 25 mg |
| For Use With (Application) | Quorum sensing, virulence gene regulation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.