Learn More
AdipoGen 2-Heptyl-3-hydroxyl-4-quinolon
SDP
Supplier:  AdipoGen CDXH0077M025
Encompass_Preferred
Chemical. CAS 108985-27-9. Formula C16H21NO2. MW 259.34. Synthetic. 2-heptyl-3-hydroxy-4-quinolone can function as an intercellular signal. Quorum sensing-regulated virulence factor used to induce and study the regulation of virulence genes such as those involved in iron scavenging. Quorum sensing is a signaling system used by bacteria to coordinate activity based upon their population density. The system involves the exchange of signaling molecules among bacteria via cell receptors.
Catalog No. 50-256-0767
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.