Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Heptylamine 97.0+%, TCI America™
SDP

Catalog No. H0645250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0645250ML 250 mL
H064525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0645250ML Supplier TCI America Supplier No. H0645250ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 123-82-0
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00008101
InChI Key VSRBKQFNFZQRBM-UHFFFAOYNA-N
Synonym 2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
PubChem CID 5603
IUPAC Name heptan-2-amine
SMILES CCCCCC(C)N

Specifications

Color Yellow
Boiling Point 142°C
Quantity 250 mL
UN Number 2734
Formula Weight 115.22
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Heptylamine
RTECSNumber MQ5425000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.