Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hexanol 98.0+%, TCI America™
SDP

Catalog No. H0131100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0131100ML 100 mL
H013125ML 25 mL
H0131500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0131100ML Supplier TCI America Supplier No. H0131100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 626-93-7
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004585
InChI Key QNVRIHYSUZMSGM-UHFFFAOYSA-N
Synonym 2-hexanol, sec-hexyl alcohol, 2-hydroxyhexane, 2-hexyl alcohol, n-butylmethylcarbinol, 2-hexanol, r, 2-hexanol, s, n-c4h9ch oh ch3, methylamyl alcohol, sec-hexanol
PubChem CID 12297
IUPAC Name hexan-2-ol
SMILES CCCCC(C)O

Specifications

Color Colorless
Boiling Point 139°C
Quantity 100 mL
UN Number 2282
Formula Weight 102.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hexanol
EINECSNumber (2)-0217
RTECSNumber MO8470000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.