Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hexanone 98.0+%, TCI America™
SDP

Catalog No. H0114100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0114100ML 100 mL
H011425ML 25 mL
H0114500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0114100ML Supplier TCI America Supplier No. H0114100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 591-78-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009482
InChI Key QQZOPKMRPOGIEB-UHFFFAOYSA-N
Synonym 2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
PubChem CID 11583
IUPAC Name hexan-2-one
SMILES CCCCC(C)=O

Specifications

Melting Point -57°C
Color Yellow
Boiling Point 127°C
Quantity 100 mL
UN Number 1224
Formula Weight 100.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hexanone
EINECSNumber (2)-0542
RTECSNumber MP1400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.