Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hexyl-1-n-octanol 98.0+%, TCI America™
SDP

Catalog No. H16351ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16351ML 1 mL
H16355ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H16351ML Supplier TCI America Supplier No. H16351ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19780-79-1
Molecular Formula C14H30O
Molecular Weight (g/mol) 214.39
MDL Number MFCD00046769
InChI Key QNMCWJOEQBZQHB-UHFFFAOYSA-N
Synonym 2-Hexyl-n-octyl Alcohol
PubChem CID 545551
IUPAC Name 2-hexyloctan-1-ol
SMILES CCCCCCC(CO)CCCCCC

Specifications

Color Yellow
Boiling Point 162°C
Quantity 1 mL
Formula Weight 214.39
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hexyl-1-n-octanol
EINECSNumber (2)-0217&(2)-3704
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.