Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hexyne 98.0+%, TCI America™
SDP

Catalog No. H042925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H042925ML 25 mL
H04295ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H042925ML Supplier TCI America Supplier No. H042925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 764-35-2
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00009470
InChI Key MELUCTCJOARQQG-UHFFFAOYSA-N
Synonym 2-hexyne, methyl propyl acetylene, methylpropylacetylene, acmc-209p3f, 1-methyl-2-propylacetylene, c3h7c.$.cch3
PubChem CID 33629
IUPAC Name hex-2-yne
SMILES CCCC#CC

Specifications

Color Yellow
Boiling Point 84°C
Quantity 25 mL
UN Number 3295
Formula Weight 82.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hexyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.